Learn More
Karl Hecht™ Assistent™ DURAN™ Round Bottom Flasks
Product Code. 10063711
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Closure Size:
29/32
NS 14/23
NS 19/26
NS 29/32
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Size unitSize
10063711 100 mL NS 14/23 Each
10155711 100 mL NS 19/26 Each
10195851 1000 mL NS 29/32 Each
10545382 2000 mL NS 29/32 Each
10628471 250 mL 29/32 Each
10629241 500 mL 29/32 Each
10165471 50 mL NS 29/32 Each
10217191 50 mL NS 19/26 Each
10277231 100 mL NS 29/32 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10063711 Supplier Karl Hecht™ Supplier No. 41913014
Only null left
Add to basket
Add to basket
For the safe and easy mixing, pouring and storage of chemicals in the laboratory. Karl Hecht™ Assistent™ DURAN™ Round Bottom Flasks are manufactured from durable DURAN borosilicate 3.3 glass.
  • Without stopper
  • With standard ground
  • Reinforced ground neck marked with standard ground size
Specifications
Capacity (Metric) 100 mL
Material Borosilicate Glass, Duran Glass
Bottom Shape Round
Closure Size NS 14/23