Learn More
Karl Hecht™ Assistent™ DURAN™ Round Bottom Flasks
Description
- Without stopper
- With standard ground
- Reinforced ground neck marked with standard ground size
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Material | Borosilicate Glass, Duran Glass |
| Bottom Shape | Round |
| Closure Size | NS 29/32 |