missing translation for 'onlineSavingsMsg'
Learn More

KARTELL DOSIPET S

Product Code. 10059782
Change view
Click to view available options
Volume (Metric):
10 mL
25 mL
50 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10059782 50 mL Each
10020263 10 mL Each
10253143 25 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10059782 Supplier KARTELL Supplier No. 8307200
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Dosipet S is a series of adjustable volume repetitive dosing devices. They are made of high quality materials ensuring considerable mechanical and chemical resistance.

  • Useful for dosing concentrated acids, polar and no-polar solvents.
  • High quality components and assembly make this series the ideal choice for measuring out organic polar and non- polar solvents, including halogenated solvents, and concentrated mineral acids, including particularly volatile acids such as nitric and trifluoroacetic acids.
  • In addition, they can be used with peroxides, hydrochloric acid and sulphuric acid.
  • It comes with a borosilicate glass piston.
  • The reagents come into contact only with parts made of PTFE
  • Precise and quick volume adjustment using the analogue slider
  • Precise graduated scale to ensure reproducibility of the dispensed volumes
  • No loss of reagent when eliminating air or filling
  • Dispensing without bubbles forming
  • The piston completely transfers the liquid with every movement
  • Total recovery of the liquid in the discharge tube by turning the delivery tip 180°
  • Autoclavable at +121°C for 20 minutes

Specifications

Accuracy ± 0.3 mL
Product Type Dosipet S
Autoclavable Autoclavable
Color White
Channels 1
Material Borosilicate Glass, PTFE
Volume (Metric) 50 mL
Display Analog
Increments 1.0 mL
Volume (English) 1.69 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.