missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Isoserine, 98%

Product Code. 10187843 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10187843 1 g Glass Bottle 1g
10026153 250 mg Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10187843 Supplier Thermo Scientific Chemicals Supplier No. 279670010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 565-71-9 | C3H7NO3 | 105.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565-71-9
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.09
MDL Number MFCD00008138
InChI Key BMYNFMYTOJXKLE-UHFFFAOYSA-N
Synonym dl-isoserine, isoserine, 3-amino-2-hydroxypropionic acid, propanoic acid, 3-amino-2-hydroxy, 2-hydroxy-3-aminopropanoic acid, h-dl-isoser-oh, inverted exclamation marka-3-amino-2-hydroxypropionic acid, rs-isoserine, pubchem23947
PubChem CID 11267
IUPAC Name 3-amino-2-hydroxypropanoic acid
SMILES C(C(C(=O)O)O)N

Specifications

Melting Point 235°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula NH2CH3CH(OH)COOH
Quantity 1 g
Formula Weight 105.09
Percent Purity 98%
Physical Form Powder
Chemical Name or Material DL-Isoserine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.