Learn More
BOLA Distributors for Canisters
Product Code. 10150225
Change view
Click to view available options
Quantity:
55
60
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10150225 60 Each
15502140 55 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10150225 Supplier BOLA Supplier No. D76024
Only null left
Add to basket
Add to basket
Distributors for Canisters PTFE. Screw cap made of glass-fibre reinforced PTFE.
  • Distributors for canisters with movable body and GL-threaded necks.
  • Screw cap made of glass-fibre reinforced PTFE
  • Body made of PTFE.
Specifications
Color White
Quantity 60
Sterility Non-sterile
Thread Size (Metric) 3 x 18 mm
Certifications/Compliance FDA
Disposable Reusable
Closure Liner Unlined
Autoclavable Autoclavable