Learn More
Lenz Laborglasintrumente Distilling Receivers Acc. To Anschütz-Thiele, Straight
Product Code. 10191661
Change view
Click to view available options
Standard Taper Joint:
Socket and Cone NS 14/23
Socket and Cone NS 29/32
Capacity (Metric):
25 mL
50 mL
100 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Capacity (Metric) unitSize
10191661 Socket and Cone NS 29/32 100 mL Each
10579401 Socket and Cone NS 14/23 25 mL Each
10129220 Socket and Cone NS 29/32 50 mL Each
10355831 Socket and Cone NS 14/23 25 mL Each
10758461 Socket and Cone NS 29/32 100 mL Each
10788271 Socket and Cone NS 29/32 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10191661 Supplier Lenz Laborglasintrumente Supplier No. 05295014
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Receivers with straight adapter, graduated, with NS socket and NS cone
  • Vacuum connection: thread GL18
  • NS stopcock with solid glass or PTFE plug
  • Made of DURAN™ tubing
Specifications
Material Duran Glass
Capacity (Metric) 100 mL
Standard Taper Joint Socket and Cone NS 29/32