Learn More
Lenz Laborglasintrumente Distilling Receivers Acc. To Anschütz-Thiele, Straight
Description
- Receivers with straight adapter, graduated, with NS socket and NS cone
- Vacuum connection: thread GL18
- NS stopcock with solid glass or PTFE plug
- Made of DURAN™ tubing
Specifications
Specifications
| Material | Duran Glass |
| Capacity (Metric) | 50 mL |
| Standard Taper Joint | Socket and Cone NS 29/32 |