missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOLA Distillation Apparata
Description
- Distillation Apparata of PTFE - with SAFE LAB-System consisting of: Reaction vessel
- flat flange (B281-03) Gasket
- flat flange (B 282-02) Lid
- flat flange (B 283-02) Joining piece
- flat flange (B 284-02) Dropping funnel
- 125 ml (B 285-01) Supporting device for thermometer (B 286-03)
- Thermometer for flasks (B 287-03)
- Stirrer bearing (B288-02)
- Moon-Shaped stirrer shaft (C 376-12)
- Stillhead 2x NS 29/32 (B 289-03)
- Distillation thermometer (B 290-03)
- Liebig condenser 270 mm (B 291-02)
- Receiver adapter (B 292-02)
- Vacuum stopcock (B 293-02)
- Round-bottom flask (A 158-06)
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Dimensions (L x W x H) | 550 x 700 mm |
| Autoclavable | Autoclavable |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?