Learn More
Kartell™ Disposable Petri Dishes
Product Code. 10061001
Change view
Click to view available options
Diameter (Metric):
90 mm
Unit Size:
Pack of 500
Product Code. Diameter (Metric) unitSize
10061001 90 mm Pack of 500
1 options
Product Code. 10061001 Supplier Kartell™ Supplier No. 0036200
Only null left
Add to basket
Add to basket
Prevent condensation with this ventilated petri dish. Kartell™ Disposable Petri Dishes are best for preventing condensation and ideal for use in routine lab work and bacteriological research.
  • With aeration notches on the lid to prevent condensation formation
  • Aseptic
  • Polystyrene
  • ß Beta-ray sterilization
Specifications
Certifications/Compliance CE Marked
Color Clear
Disposable Single-use
Material Polystyrene
Shape Round
Sterility Sterile
Vent No
Diameter (Metric) 90 mm