Learn More
Greiner Bio-One Disposable Inoculating Loops
Product Code. 10194711
Change view
Click to view available options
Volume:
1 μL
10 μL
Color:
Blue
White
Packaging Type:
10/bag
Unit Size:
Pack of 3000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume Color Packaging Type unitSize
10194711 1 μL White 10/bag Pack of 3000
10240921 10 μL Blue 10/bag Pack of 3000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10194711 Supplier Greiner Bio-One Supplier No. 731165
Only null left
Add to basket
Add to basket
For use in picking single colonies. Greiner Bio-One Disposable Inoculating Loops are flexible for collection and inoculation.
  • Sterilised by irradiation
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
Volume 1 μL
Length (Metric) 200 mm
Material Polystyrene
Sterility Sterile
Color White
Quantity 3000
Packaging Type 10/bag