Learn More
Hirschmann™ Discharge Tube Unit with Luer-Lock Connection
Description
- Guided tubing
- Screw-coupled
- With Luer-Lock connector for filtration
Specifications
Specifications
| Quantity | 1 Each |
| Type | Ejection Tube Unit |
| For Use With (Equipment) | ceramus™ classic and EM-dispenser for 30 + 60 mL |
| Material | FEP, PFA |
| Description | With Luer-Lock-connection |