missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl itaconate, 97%, stabilized

Product Code. p-4510128 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10061600 100 g Glass bottle 100g
10143340 500 g Glass bottle 500g
11975031 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10061600 Supplier Thermo Scientific Acros Supplier No. 149161000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 617-52-7 | C7H10O4 | 158.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-52-7
Molecular Formula C7H10O4
Molecular Weight (g/mol) 158.15
MDL Number MFCD00008446
InChI Key ZWWQRMFIZFPUAA-UHFFFAOYSA-N
Synonym dimethyl itaconate, dimethyl 2-methylenesuccinate, dimethylitaconate, itaconic acid dimethyl ester, itaconic acid, dimethyl ester, butanedioic acid, methylene-, dimethyl ester, 1,4-dimethyl 2-methylidenebutanedioate, dimethyl methylenesuccinate, unii-11jib0yi93, succinic acid, methylene-, dimethyl ester
PubChem CID 69240
SMILES COC(=O)CC(=C)C(=O)OC

Specifications

Melting Point 34.0°C to 39.0°C
Color White
Density 1.2400g/mL
Boiling Point 208.0°C
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3O2CCH2C(=CH2)CO2CH3
Quantity 100 g
Beilstein 02,762
Specific Gravity 1.24
Solubility Information Solubility in water: slightly soluble
IUPAC Name dimethyl 2-methylidenebutanedioate
Formula Weight 158.15
Percent Purity 97%
Physical Form Crystalline Low Melting Mass or Liquid
Chemical Name or Material Dimethyl itaconate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.