missing translation for 'onlineSavingsMsg'
Learn More

Diisopropylamine, 99+%, Thermo Scientific Chemicals

Product Code. 10444715 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic Drum
Unit Size:
1 L
250mL
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10444715 5 L Plastic Drum 5 L
10092890 250 mL Glass bottle 250mL
10434335 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10444715 Supplier Thermo Scientific Acros Supplier No. 219430051
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-18-9
Molecular Weight (g/mol) 101.19
MDL Number MFCD00008862
InChI Key UAOMVDZJSHZZME-UHFFFAOYSA-N
Synonym dipa, 2-propanamine, n-1-methylethyl, di-isopropylamine, n-1-methylethyl-2-propanamine, n-isopropyl-1-amino-2-methylethane, diisopropylamine, n-isopropylpropan-2-amine, n,n-diisopropylamine, bis propan-2-yl amine, diisopropyl amine
PubChem CID 7912
SMILES CC(C)NC(C)C

Specifications

Melting Point -61.0°C
Density 0.7160 g/mL
Boiling Point 84.0°C
Flash Point -17°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic Drum
Molecular Formula C6H15N
Linear Formula [(CH3)2CH]2NH
Refractive Index 1.3910 to 1.3930
Quantity 5 L
Beilstein 04, 154
Merck Index 15, 3215
Specific Gravity 0.716
Solubility Information Solubility aq. soln.: 100 g/L (20°C). Other solubilities: soluble in alcohol,soluble in most common organic solvents
IUPAC Name bis(propan-2-yl)amine
Viscosity 0.4 mPa.s (20°C)
Formula Weight 101.19
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Diisopropylamine, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.