missing translation for 'onlineSavingsMsg'
Learn More

Diflunisal Methyl Ester, Mikromol™

Product Code. 30049954
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Product Code. Quantity unitSize
30049954 25 mg 25mg
30049642 100 mg 100mg
30458666 4 x 25 mg 100mg
3 options
This item is not returnable. View return policy

Product Code. 30049954

Brand: Mikromol MM3050.010025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Diflunisal Methyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diflunisal Methyl Ester
CAS 55544-00-8
Analyte or Component Names Diflunisal Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C14 H10 F2 O3
Certifications/Compliance ISO 17025
Synonym Diflunisal Methyl Ester, 2',4'-Difluoro-4-hydroxy-[1,1'-biphenyl]-3-carboxylic acid methyl ester, Diflunisal methyl ester, Methyl 2',4'-difluoro-4-hydroxybiphenyl-3-carboxylate, Methyl 5-(2,4-difluorophenyl)salicylate
InChI Formula InChI=1S/C14H10F2O3/c1-19-14(18)11-6-8(2-5-13(11)17)10-4-3-9(15)7-12(10)16/h2-7,17H,1H3
SMILES COC(=O)c1cc(ccc1O)c2ccc(F)cc2F
IUPAC Name methyl 5-(2,4-difluorophenyl)-2-hydroxybenzoate
Molecular Weight (g/mol) 264.22
Quantity 25 mg
Formula Weight 264.0598 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.