missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodiphenylsilane, 97%, AcroSeal™

Product Code. 10043957 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10043957 100 mL AcroSeal™ Glass Bottle 100mL
10053957 800 mL AcroSeal™ Glass Bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10043957 Supplier Thermo Scientific Acros Supplier No. 430891000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 80-10-4 | C12H10Cl2Si | 253.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-10-4
Molecular Formula C12H10Cl2Si
Molecular Weight (g/mol) 253.20
MDL Number MFCD00000489
InChI Key OSXYHAQZDCICNX-UHFFFAOYSA-N
Synonym diphenyldichlorosilane, silane, dichlorodiphenyl, dichloro diphenyl silane, diphenylsilicon dichloride, diphenylsilyl dichloride, dichlor-difenylsilan, diphenyl dichlorosilane, dichlor-difenylsilan czech, hsdb 316, benzene, 1,1'-dichlorosilylene bis
PubChem CID 6627
IUPAC Name dichloro(diphenyl)silane
SMILES Cl[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point -22.0°C
Color Colorless to Yellow
Density 1.2040g/mL
Boiling Point 305.0°C
Flash Point 155°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula (C6H5)2SiCl2
Refractive Index 1.5765 to 1.5815
Quantity 100 mL
Specific Gravity 1.204
Solubility Information Solubility in water: decomposes
Formula Weight 253.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dichlorophenylsilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dichlorodiphenylsilane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.