missing translation for 'onlineSavingsMsg'
Learn More

Diacetic Aceclofenac, TRC

Product Code. 30361211
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30361211 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30361211 Supplier Toronto Research Chemicals Supplier No. TRCD3045305MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diacetic Aceclofenac
CAS 1216495-92-9
Molecular Formula C20 H17 Cl2 N O8
Quantity 5 mg
Synonym [[[[[[[2-[(2,6-Dichlorophenyl)amino]phenyl]acetyl]oxy]acetyl]oxy]acetyl]oxy]acetic Acid, Aceclofenac Imp. H (EP), Diacetic Aceclofenac, 2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid 2-[2-(carboxymethoxy)-2-oxoethoxy]-2-oxoethyl ester
InChI Formula InChI=1S/C20H17Cl2NO8/c21-13-5-3-6-14(22)20(13)23-15-7-2-1-4-12(15)8-17(26)30-10-19(28)31-11-18(27)29-9-16(24)25/h1-7,23H,8-11H2,(H,24,25)
SMILES OC(=O)COC(=O)COC(=O)COC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl
IUPAC Name 2-[2-[2-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxyacetyl]oxyacetyl]oxyacetic acid
Molecular Weight (g/mol) 470.26
Formula Weight 469.0331
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.