missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ DexPure Nitrile Gloves, Powder-Free

Product Code. 12676103
Click to view available options
Size:
10
9
Large
Small
Unit Size:
Pack of 100
Pack of 50
This item is not returnable. View return policy

Product Code. 12676103

Brand: Honeywell Safety Products™ 4580091S

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Certified for food contact and resistant to oils, fats and detergents, these powder-free, 100% nitrile gloves are a good alternative to latex. Honeywell Safety Products™ DexPure Nitrile Gloves, Powder-Free have improved grip and are treated to prevent bacteria.

  • Nitrile construction for chemical and mechanical resistance.
  • Patterned finish for good grip.
  • Certified for food contact and treated to prevent bacteria.
  • Suitable for the following:
    • Agrifood industry.
    • Electronics and complex assembly.
    • Graphic industry.
    • Painting in the automotive industry.
    • Pharmaceutical laboratories.
    • Chemical handling.
  • Lightly powdered to facilitate donning and doffing.
  • Single use.
  • Alternative for latex allergy sufferers.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standards, EN 374-2003 Chemical hazards, EN 455-1&2 Medical Standards, European directive N°1934/2004:Food contact, C Category PPE 2, EC Certificate Number 0075/797/162/12/08/0490, EC Attestation Number 0075/797/162/12/08/0490
Classification Dexterity: 5
Color Blue
Disposable Single-use
Exterior Finish Patterned
Material Nitrile
Powdered Powdered
Product Type Gloves
Size Small
Thickness (Metric) 0.1 mm
For Use With (Application) Automobile, Aviation, Catering, Chemical, Food, Industrial Cleaning, Laboratory, Maintenance, Medical, Pharmaceutical, Petrochemical, Printing and Water Treatment industries.
Quantity 100/PK
Resistant To Chemicals, Mechanical Stress
Length (Metric) 24 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.