missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Desmocollin 2/3 Monoclonal Antibody (7G6)

Description
Desmocollin 2/3 Monoclonal Antibody for IHC (P), IP, Western Blot
This gene encodes a member of the desmocollin protein subfamily. Desmocollins, along with desmogleins, are cadherin-like transmembrane glycoproteins that are major components of the desmosome. Desmosomes are cell-cell junctions that help resist shearing forces and are found in high concentrations in cells subject to mechanical stress. This gene is found in a cluster with other desmocollin family members on chromosome 18. Mutations in this gene are associated with arrhythmogenic right ventricular dysplasia-11, and reduced protein expression has been described in several types of cancer. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
| Antigen | Desmocollin 2/3 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 7G6 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.4 |
| Gene | DSC3 |
| Gene Accession No. | Q02487, Q14574 |
| Gene Alias | ARVC10; ARVD10; ARVD11; cadherin family member 2; Cadherin family member 3; CDHF2; CDHF3; CDHF5; CDHF6; CMD1BB; desmocollin 2; desmocollin 3; desmocollin-2; Desmocollin-3; desmocollin-4; desmoglein-2; desmoglein-3; Desmosomal glycoprotein II; desmosomal glycoprotein II/III; Desmosomal glycoprotein III; DG2; DGII/III; DSC; DSC1; DSC2; DSC3; DSC4; HDGC; HT-CP; PVA |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?