missing translation for 'onlineSavingsMsg'
Learn More

Desethyl Terbuthylazine-d9, TRC

Product Code. 30365767
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365767 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30365767 Supplier Toronto Research Chemicals Supplier No. TRCD28970210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Desethyl Terbuthylazine-d9
CAS 1219798-52-3
Molecular Formula C7 2H9 H3 Cl N5
Quantity 10 mg
Synonym Terbuthylazine-desethyl D9 (tert-butyl D9), N-tert-Butyl-6-chloro-1,3,5-triazine-2,4-diamine D9 (tert-butyl D9)
InChI Formula InChI=1S/C7H12ClN5/c1-7(2,3)13-6-11-4(8)10-5(9)12-6/h1-3H3,(H3,9,10,11,12,13)/i1D3,2D3,3D3
SMILES [2H]C([2H])([2H])C(Nc1nc(N)nc(Cl)n1)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC Name 6-chloro-4-N-[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]-1,3,5-triazine-2,4-diamine
Molecular Weight (g/mol) 210.71
Formula Weight 210.1346
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.