missing translation for 'onlineSavingsMsg'
Learn More

Decon™ Decon 90

Product Code. 10053113
Change view
Click to view available options
Volume (Metric):
1000 mL
5000 mL
20 L
Unit Size:
1 L
20 L
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10053113 20 L 20 L
10335650 1000 mL 1 L
10549760 5000 mL 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10053113 Supplier Decon™ Supplier No. D9020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Biodegradable, phosphate and enzyme free, water rinsable surface active cleaning agent and radioactive decontaminant

  • Phosphate and enzyme free
  • A 2% to 5% solution is adequate for the majority of cleaning applications
  • Increasing solution strength, heating the solution or use in an ultrasonic cleaner will reduce the cleaning time
  • Not suitable for use on non-ferrous metals, notably aluminum and zinc, or on polycarbonate
  • Concentrate
  • pH 13.0

Specifications

Product Type Liquid Detergent
pH 13.0 (undiluted), 11.9 (10%), 10.7 (2%)
Appearance Milky white
Boiling Point 100.5°C
Composition An emulsion of anionic and non-ionic surface active agents, stabilizing agents, non-phosphate deterg
Container Type Polyethylene containers
Volume (Metric) 20 L
compliance-icons
Product Identifier
  • DECON 90
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Supplemental information
  • MIXTURE LIST-Contain: Potassium Hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.