missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dansyl-d6 Chloride (N,N-dimethyl-d6), CDN
Specifications
Specifications
| Chemical Name or Material | Dansyl Chloride-d6 |
| CAS | 1276379-68-0 |
| Molecular Formula | C12 2H6 H6 Cl N O2 S |
| Quantity | 0.05 g |
| InChI Formula | InChI=1 S/C12H12ClNO2S/c1-14(2)11-7-3-6-10-9(11)5-4-8-12(10)17(13,15)16/h3-8 H,1-2H3/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])N(c1cccc2c(cccc12)S(=O)(=O)Cl)C([2 H])([2 H])[2 H] |
| IUPAC Name | 5-[bis(trideuteriomethyl)amino]naphthalene-1-sulfonyl chloride |
| Molecular Weight (g/mol) | 275.78 |
| Formula Weight | 275.0654 g/mol |
| Percent Purity | 99 atom % D, min 95% Chemical Purity |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?