missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dansyl Chloride-d6, TRC
Specifications
Specifications
| CAS | 1276379-68-0 |
| Molecular Formula | C12 2H6 H6 Cl N O2 S |
| Quantity | 10 mg |
| Synonym | 1-Naphthalenesulfonyl chloride, 5-[di(methyl-d3)amino]- (ACI), 5-[Di(methyl-d3)amino]-1-naphthalenesulfonyl chloride (ACI), Dansyl Chloride-d6, Dansyl-d6 Chloride (N,N-dimethyl-d6), Dansyl Chloride-D6 (N,N-dimethyl-D6) |
| InChI Formula | InChI=1S/C12H12ClNO2S/c1-14(2)11-7-3-6-10-9(11)5-4-8-12(10)17(13,15)16/h3-8H,1-2H3/i1D3,2D3 |
| SMILES | [2H]C([2H])([2H])N(c1cccc2c(cccc12)S(=O)(=O)Cl)C([2H])([2H])[2H] |
| IUPAC Name | 5-[bis(trideuteriomethyl)amino]naphthalene-1-sulfonyl chloride |
| Molecular Weight (g/mol) | 275.78 |
| Formula Weight | 275.0654 |
| Chemical Name or Material | Dansyl Chloride-d6 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?