Learn More
Specac™ Cyclone™ C5 Long Pathlength Gas Cell MCFV
Product Code. 15270278
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
1.33 L
Product Code. Volume (Metric)
15270278 1.33 L
1 options
Product Code. 15270278 Supplier Specac™ Supplier No. GS24105MCFV
This is a heatable long pathlength gas cell with metal body, CaF2 windows, fixed pathlength and Viton O-rings. The cell has a volume of 1.33 litres.
Specifications
Description Fixed pathlength and Viton O-rings
Volume (Metric) 1.33 L