Learn More
Invitrogen™ Custom RNA, 5' Biotin
Product Code. 10143713
Change view
Click to view available options
Product Type:
RNA 5' Bio
RNA 5' Bio In-vivo
Quantity:
20 nmol
5 nmol
500 nmol
Unit Size:
20nmol
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity unitSize
10143713 RNA 5' Bio In-vivo 5 nmol Each
10552364 RNA 5' Bio 20 nmol 20nmol
10638283 RNA 5' Bio In-vivo 20 nmol Each
10134403 RNA 5' Bio In-vivo 500 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10143713 Supplier Invitrogen™ Supplier No. 100001857
Custom RNA, 5' Biotin
Use this tool for fast and easy online ordering of bench-tested Validated Stealth RNAi™ siRNA, ready-to-order Stealth Select RNAi™ siRNA, bench-tested BLOCK-iT™ Pol II miR Validated miRNA Vector Duopaks, and ready-to-clone BLOCK-iT™ miR RNAi Select.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Room temperature
Description RNA 5' Bio In-vivo, 5nmol, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation
Product Type RNA 5' Bio In-vivo
Quantity 5 nmol