Learn More
Invitrogen™ Custom Primer, Mixing Primers (Pooling)
Product Code. 10174092
For Research Use Only. All usage must comply with product instructions.
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10174092 Supplier Invitrogen™ Supplier No. 10668077
For Research Use Only. All usage must comply with product instructions.
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Custom Primer, Mixing Primers (Pooling)
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Description Mixing Primers, 25 nmol
Product Type Mixing Primers
Content And Storage Room Temperature
Quantity 25 nmol