Learn More
Invitrogen™ Custom Primer
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
Specifications
Specifications
| Description | HPLC Primer, 10μmol |
| Product Type | HPLC Primer |
| Quantity | 10 μmol |
| Grade | HPLC |