missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cullin 1 Monoclonal Antibody (2H4C9)
GREENER_CHOICE

Product Code. 10109112
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10109112 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10109112 Supplier Invitrogen™ Supplier No. 322400

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Mouse Monoclonal Antibody

CUL-1 Antibody (2H4C9), Prod# 32-2400 specifically detects neddylated and unmodified Cullin 1 in human cells.

CUL1 gene ontology annotations related to this gene include animal organ morphogenesis; apoptotic process; cell proliferation; protein monoubiquitination; SCF-dependent proteasomal ubiquitin-dependent protein catabolic process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cullin 1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2H4C9
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene CUL1
Gene Accession No. Q13616
Gene Alias Cul1; CUL-1; cullin 1; cullin-1
Gene Symbols CUL1
Host Species Mouse
Immunogen A synthetic peptide corresponding to an 11 amino acid sequence at the C-terminus of the human CUL-1 protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8454
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.