missing translation for 'onlineSavingsMsg'
Learn More

CRAC intermediate 1, MedChemExpress

Product Code. p-200096449
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16450478 5 mg 5mg
16448018 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16450478 Supplier Medchem Express Supplier No. HY20587

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CRAC intermediate 1 is a key intermediate in the chemical synthesis of a series of CRAC channel inhibitors, detailed information can be found in Patent WO 2010122089 A1, intermediate 9.

CRAC intermediate 1 is a key intermediate in the chemical synthesis of a series of CRAC channel inhibitors, detailed information can be found in Patent WO 2010122089 A1, intermediate 9.

Specifications

Chemical Name or Material CRAC intermediate 1
CAS 1249343-86-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C10H7F2N3O
Quantity 5 mg
Solubility Information DMSO : ≥ 50 mg/mL (224.03 mM)
SMILES O=C(C1=C(F)C=CC=C1F)NC2=NNC=C2
Molecular Weight (g/mol) 223.18
Formula Weight 223.18
Percent Purity 98.46%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.