missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRAC intermediate 1, MedChemExpress
Description
CRAC intermediate 1 is a key intermediate in the chemical synthesis of a series of CRAC channel inhibitors, detailed information can be found in Patent WO 2010122089 A1, intermediate 9.
Specifications
Specifications
| Chemical Name or Material | CRAC intermediate 1 |
| CAS | 1249343-86-9 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C10H7F2N3O |
| Quantity | 5 mg |
| Solubility Information | DMSO : ≥ 50 mg/mL (224.03 mM) |
| SMILES | O=C(C1=C(F)C=CC=C1F)NC2=NNC=C2 |
| Molecular Weight (g/mol) | 223.18 |
| Formula Weight | 223.18 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?