Learn More
CR Tubing for Julabo Cryo-Compact Circulator
Product Code. 10112011
Change view
Click to view available options
For Use With (Equipment):
Models CF30, CF31, CF40, CF41, ED
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
10112011 Models CF30, CF31, CF40, CF41, ED Each
1 options
Product Code. 10112011 Supplier Julabo Supplier No. 8930008
Only null left
Add to basket
Add to basket
Specifications
Description CR
For Use With (Equipment) Models CF30, CF31, CF40, CF41, ED
For Use With (Application) Ideal for temperature application to external systems
Length (Metric) 1 m
Diameter (Metric) 8 mm