Learn More
CPC (Colder) AseptiQuik™ S Low-Flow Genderless Quick-Disconnect Fittings, Sanitary Adapter, Polycarbonate
Product Code. 30054369
Change view
Click to view available options
Includes:
1/4 in. Sanitary Clamp Size
3/4 in. Sanitary Clamp Size
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
30054369 3/4 in. Sanitary Clamp Size Each
30054260 1/4 in. Sanitary Clamp Size Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30054369 Supplier CPC (Colder) Supplier No. 5011201
Only null left
Add to basket
Add to basket
Low-flow genderless sterile connectors simplify process integration while maximizing flexibility
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants. Simple flip, click, and pull design minimizes operator errors. Create quick and easy sterile connections, even in nonsterile environments. Specifically designed for small-volume fluid transfer applications. Robust construction provides reliable and repeatable fluid transfer. 0.25 in. nominal flow path size
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Material Tygon 2475
Min. Operating Temperature (Metric) 1°C
Outlet Port Quick Disconnect x Sanitary
Sterilization Method 130°C
Shape Straight
Includes 3/4 in. Sanitary Clamp Size
Material (Seal) Platinum-cured Silicone
Color White with white pull tab
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.