missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Composed of 96 polypropylene tubes in a standard microplate format.
- Features 1.2 mL tubes that are available individually or in strips of eight tubes.
- Polyethylene tube caps are available in 8-cap strips.
Specifications
Specifications
| Product Type | 8-cap Strip |
| Material | Polyethylene |
| Color | Clear |
| No. per Pack | 12/Pk. |
| No. per Case | 120/Cs. |
| Sterility | Sterile |
| For Use With (Equipment) | 1.2 mL 98-well Cluster Tube System |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?