Description
A stacking device models the top surface of the CellSTACK™ chamber, securely nesting in the strengthening ribs. It is used to optimize incubator space while providing clearance for venting
Specifications
Specifications
| For Use With (Equipment) | Incubator |
| Material | Plastic |
| Type | Stacker |
| For Use With (Application) | Incubator |