missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Specifications
| Melting Point | 474.0°C |
| Color | Beige-White to Green |
| Physical Form | Powder |
| Quantity | 1 kg |
| Assay Percent Range | 98.5% min. (ex Cu) |
| Linear Formula | CuCN |
| Fieser | 05,166; 06,146; 09,124; 16,97 |
| Merck Index | 15, 2651 |
| Solubility Information | Solubility in water: practically insoluble in water. Other solubilities: soluble in acids and alkalies, practically insoluble in alcohol |
| Formula Weight | 89.56 |
| Show More |
Safety and Handling
Product Identifier
- Copper(I) cyanide
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- H310-Fatal in contact with skin.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H330-Fatal if inhaled.
- H300-Fatal if swallowed.
- EUH032-Contact with acids liberates very toxic gas.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?