missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) bromide, 98%, extra pure

Product Code. 10193830 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10193830 1 kg Glass bottle 1kg
10072520 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10193830 Supplier Thermo Scientific Acros Supplier No. 199580010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7787-70-4 | BrCu | 143.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-70-4
Molecular Formula BrCu
Molecular Weight (g/mol) 143.45
MDL Number MFCD00010969
InChI Key NKNDPYCGAZPOFS-UHFFFAOYSA-M
Synonym copper i bromide, copper bromide cubr, copper 1+ bromide, copper monobromide, cuprousbromide, hsdb 270, cubr, copper 1 bromide
PubChem CID 24593
IUPAC Name bromocopper
SMILES [Cu+].[Br-]

Specifications

Boiling Point 1345.0°C
Melting Point 504.0°C
Color Green
Physical Form Powder
Quantity 1 kg
Assay Percent Range 97.5% min. (Complexometry)
Linear Formula CuBr
Fieser 01,165; 02,90; 03,67; 04,108; 05,163; 06,143; 07,79; 08,116; 11,140; 12,286; 14,99; 16,96
Merck Index 15, 2649
Solubility Information Solubility in water: soluble in cold water. Other solubilities: decomposes in hot water, sol. in hcl, hbr, nh4oh (formation of complexes), prectically insoluble in conc. h2so4 and acetone, insoluble matter in hcl: ≤0.02%
Formula Weight 143.45
Percent Purity 98%
Grade Extra Pure
Packaging Glass bottle
Specific Gravity 4.71
Density 4.7100g/mL
Chemical Name or Material Copper(I) bromide
Show More Show Less
compliance-icons
Product Identifier
  • Copper(I) bromide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.