Description
Extended controlled dropper tip dispenses one drop at a time
Specifications
Specifications
| Color | White |
| Material | LDPE |
| Quantity | 1000Pack |
| Thread Size (Metric) | 24 mm |
| Neck Finish | 24 mm |
| Drop Size | 41 μL |
| Product Type | Dropper Tip |