Learn More
HUBER Control Panel Extension Cable for Pilot ONE
Product Code. 10195253
Change view
Click to view available options
For Use With (Equipment):
Pilot One
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
10195253 Pilot One Each
1 options
Product Code. 10195253 Supplier HUBER Supplier No. 16160
Only null left
Add to basket
Add to basket
Control panel extension cable for Pilot ONE for using the controller as remote control (without RS232)
  • Control panel extension cable
  • Unistat pilot - Unistat control
  • CC-Pilot - CC/UC
  • Length: 3m
Specifications
For Use With (Equipment) Pilot One
Type Control Panel Extension Cable