missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Connexin 46 Polyclonal Antibody
GREENER_CHOICE

Product Code. 10169102
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10169102 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10169102 Supplier Invitrogen™ Supplier No. 388300

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Rabbit Polyclonal Antibody

38-8300 detects Cx46 in IHC application in cryostat sections of adult mouse lens.

Connexin 46 (Cx46), also known as Gap Junction Protein Alpha-3 (GJA3), CAE3, CX46 and C2P3, maps to human chromosome 13q11-q12 and encodes a 46 kDa protein. Cx46, along with Cx50, is principally expressed in the lens of the eye. Cx46 mediates intercellular interactions during development and is necessary for the survival of neural crest cells. Cx46 forms gap junctions that connect lens fiber cells and are crucial for maintaining lens transparency and normal lens function. Mutations of Cx46 result in severe cataracts of the lens. Individual knockouts of Cx46 and Cx50 lead to changes in the rate of lens fiber cell differentiation and cell size, thus the interaction of Cx46 and Cx50 is required for proper organization of fiber cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 46
Applications Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene GJA3
Gene Accession No. Q64448
Gene Alias alpha 3 connexin; Cnx46; connexin 46; connexin-46; CTRCT14; Cx43; CX46; Cxn-46; CZP3; gap junction alpha 3; gap junction alpha-3 protein; gap junction membrane channel protein alpha 3; gap junction protein alpha 3; gap junction protein, alpha 3; gap junction protein, alpha 3, 46kDa; Gja3; Gja-3
Gene Symbols GJA3
Host Species Rabbit
Immunogen Synthetic peptide derived from an internal region of the mouse Connexin 46 protein.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14611
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.