missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Connexin 40 Polyclonal Antibody
GREENER_CHOICE

Product Code. 10149632
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10149632 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10149632 Supplier Invitrogen™ Supplier No. 365000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Rabbit Polyclonal Antibody

Connexin 40 Polyclonal Antibody for Western Blot, IHC (F)

Connexin 40 (Cx40), belongs to the connexin family of gap junction proteins, which are expressed in pancreas, kidney, skeletal muscle, liver, placenta and heart. Connexin 40 is highly expressed in vascular endothelial cells, and the mammalian heart (specifically in the atrial myocardium, and the His-Purkinje conduction system). At least three other connexins are expressed in the heart, including Cx37 (endocardium), Cx43 (atrial and ventricular myocardium, and distal parts of the conduction system) and Cx45 (throughout the heart). Cx40 and Cx43 frequently co-localize in cells of the cardiovascular system and participate in the formation of heterotypic gap junction channels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 40
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Gja5
Gene Accession No. Q01231
Gene Alias 5730555N10Rik; ATFB11; Cnx40; connexin 40; connexin-40; Cx40; Cxn-40; Gap junction alpha-5 protein; gap junction membrane channel protein alpha 5; gap junction protein; gap junction protein alpha 5; gap junction protein alpha 5 40kDa; gap junction protein, alpha 5; GJA5; Gja-5
Gene Symbols Gja5
Host Species Rabbit
Immunogen Synthetic peptide derived from an internal region of the mouse Connexin 40 (Cx40) protein.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14613
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.