missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Connexin 37 Polyclonal Antibody

Description
40-4300 was used in the immunofluorescence (IHC) analysis to successfully detect Connexin 37 in mouse heart sections.
This gene encodes a member of the connexin gene family. The encoded protein is a component of gap junctions, which are composed of arrays of intercellular channels that provide a route for the diffusion of low molecular weight materials from cell to cell. Mutations in this gene have been associated with atherosclerosis and a higher risk of myocardial infarction.
Specifications
Specifications
| Antigen | Connexin 37 |
| Applications | Immunohistochemistry (Frozen), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.4 |
| Gene | GJA4 |
| Gene Accession No. | P28235 |
| Gene Alias | AU020209; AW558810; Cnx37; connexin 37; connexin-37; CX37; CXN37; Cxn-37; gap junction alpha-4 protein; gap junction membrane channel protein alpha 4; Gap junction membrane channel, protein alpha 4 (connexin 37); gap junction protein alpha 4; gap junction protein, alpha 4; gap junction protein, alpha 4, 37kDa; GJA4; Gja-4 |
| Gene Symbols | GJA4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?