missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Connexin 36 Polyclonal Antibody
GREENER_CHOICE

Product Code. 10159392
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10159392 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10159392 Supplier Invitrogen™ Supplier No. 516200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Rabbit Polyclonal Antibody

Connexin 36 Polyclonal Antibody for Western Blot, ICC/IF, IHC, ELISA

Connexin-36 (CX36), is a member of the connexin gene family that is expressed predominantly in mammalian neurons. Connexins associate in groups of 6 and are organized radially around a central pore to form connexons. Each gap junction intercellular channel is formed by the conjunction of 2 connexons. Mutations in connexin-encoding genes can lead to functional and developmental abnormalities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 36
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene GJD2
Gene Accession No. O54851, O70610, Q9UKL4
Gene Alias connexin 36; connexin36; connexin-36; cx36; Gap junction alpha-9 protein; gap junction delta-2 protein; gap junction membrane channel protein alpha 9; gap junction protein delta 2; gap junction protein, alpha 9; gap junction protein, delta 2; gap junction protein, delta 2, 36kDa; GJA10; Gja9; GJD2
Gene Symbols GJD2
Host Species Rabbit
Immunogen Peptide corresponding to a sequence located in the cytoplasmic loop between the second and third transmembrane domains of rat and mouse Connexin 36 (Cx36). This sequence differs from the human Cx36 protein by a single amino acid.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14617, 50564, 57369
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.