missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Connexin 32 Polyclonal Antibody
GREENER_CHOICE

Product Code. 10038112
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10038112 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10038112 Supplier Invitrogen™ Supplier No. 710600

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Rabbit Polyclonal Antibody

71-0600 was used in the immunofluorescence (IHC) and western blot analysis to successfully detect Connexin 32 in mouse liver.

This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 32
Applications ELISA, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene GJB1
Gene Accession No. P08033, P08034, P28230
Gene Alias CMTX; CMTX1; Cnx32; connexin 32; connexin-32; CX32; Cxn-32; GAP junction 28 kDa liver protein; gap junction beta-1 protein; gap junction membrane channel protein beta 1; gap junction protein beta 1; gap junction protein, beta 1; gap junction protein, beta 1, 32kDa; Gjb1
Gene Symbols GJB1
Host Species Rabbit
Immunogen Peptide corresponding to a portion of the cytoplasmic loop of rat connexin 32.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14618, 2705, 29584
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.