Learn More
Lenz Laborglasintrumente Cones with Short Tip
Description
- Conical ground cones
- Made of DURAN™ tubing
Specifications
Specifications
| Joint Size | NS 24/29 |
| Thickness (Metric) Wall | 1.8 mm |
| Diameter (Metric) Outer Tube | 22 mm |
| Material | Borosilicate Glass (3.3) |