Learn More
JULABO™ Complete Carrier Trays with Test Tube Rack
Description
- Tray with 4 tube racks
- Metal construction
- Withstands temperatures up to 80 °C
- For use with JULABO™ shaking water baths SW22 / SW23
Specifications
Specifications
| For Use With (Equipment) | SW22, SW23 shaking water bath |
| Type | Carrier Tray |
| Holds | 240 Tubes, 100 x 16/17 mm dia. |
| Includes | 4 Racks |
Product Title
Haben Sie Verbesserungsvorschläge?