missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Forehead Comfort Bands for use with 3M™ Versaflo™ S-Series Headcovers and Hoods
Description
- Product Type: Forehead Comfort Band
- Description: Standard Format, CE-Approved
- Color: Grey
- For Use With (Equipment): 3M™ Versaflo™ S-Series Headcovers and Hoods
- For Use With (Application): Respiratory Protection
Specifications
Specifications
| Description | Standard, CE-Approved |
| Color | Gray |
| Disposable | Reusable |
| Product Type | Comfort Band |
| Size | Large |
| For Use With (Application) | Respiratory Protection |
| For Use With (Equipment) | Versaflo S-Series Headcovers and Hoods |
| Model | S-957L |
| Quantity | 20 Pk. |
| Sterility | Non-sterile |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?