Learn More
Lenz Laborglasintrumente Columns Acc. to Vigreux
Description
- With NS socket and NS cone
- Without glass jacket
- Horizontal and lateral indentations
- Made of DURAN™ tubing
Specifications
Specifications
| Length (Metric) | 600 mm |
| Material | Borosilicate Glass 3.3 |
| Standard Taper Joint | NS 29/32 |