Learn More
Lenz Laborglasintrumente Columns Acc. to Hempel
Description
- Columns acc. to Hempel
- With removable glass jacket
- For packings like Raschig rings or glass beads
- With NS socket and NS cone
- With removable glass jacket
- Made of DURAN™ tubing
Specifications
Specifications
| Length (Metric) | 300 mm |
| Standard Taper Joint | Cone and Socket NS 14/23 |