Learn More
Lenz Laborglasintrumente Columns Acc. to Hempel
Product Code. 10141761
Change view
Click to view available options
Standard Taper Joint:
Cone and Socket NS 14/23
Cone and Socket NS 24/29
Cone and Socket NS 29/32
Length (Metric):
300 mm
500 mm
800 mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Length (Metric) unitSize
10141761 Cone and Socket NS 29/32 300 mm Each
10020501 Cone and Socket NS 24/29 800 mm Each
10129310 Cone and Socket NS 14/23 300 mm Each
10224581 Cone and Socket NS 24/29 300 mm Each
10509581 Cone and Socket NS 24/29 500 mm Each
10749031 Cone and Socket NS 29/32 800 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10141761 Supplier Lenz Laborglasintrumente Supplier No. 05115304
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Columns acc. to Hempel
  • With removable glass jacket
  • For packings like Raschig rings or glass beads
  • With NS socket and NS cone
  • With removable glass jacket
  • Made of DURAN™ tubing
Specifications
Length (Metric) 300 mm
Standard Taper Joint Cone and Socket NS 29/32