Learn More
Invitrogen™ Collagen I Polyclonal Antibody

Description
This product requires antigen retrieval using 1 mg/mL pepsin in 0.5M acetic acid for 2 hours at 37°C and multiple buffer washes prior to staining on paraffin sections. Reconstitute with 0.5 mL of distilled water. Prior to reconstitution, store at 4°C. After reconstitution, store undiluted at -20°C, avoiding freeze/thaw cycles. For long term storage the addition of 1% BSA and 0.09% sodium azide is recommended. Rabbit anti Human collagen I antibody recognizes human Collagen I, reacting with both native and heat denatured proteins, and has been cross-absorbed against Collagen types III, IV and V.
Specifications
Specifications
| Antigen | Collagen I |
| Applications | ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | 0.01M sodium borate with 1% dextran, 1% mannitol, 0.1M NaCl and no preservative |
| Gene | COL1A2 |
| Gene Accession No. | P02452, P08123 |
| Gene Alias | AA960264; AI325291; alpha 1 type I collagen; alpha 2 type I collagen; alpha 2 type I procollagen; alpha 2(I) collagen; alpha 2(I) procollagen; alpha 2(I)-collagen; alpha1 chain of type I collagen; alpha-1 collagen (I); alpha-1 type 1 collagen; alpha-1 type I collagen; alpha-1 type-1 collagen; alpha1(I) procollagen; alpha2 chain of type I collagen; alpha-2 type I collagen; alpha-2(1) collagen; alpha2(I) collagen; COL1; Col1a1; Col1a-1; COL1A1; matrix protein; Col1a2; Col1a-2; COLA; Cola1; Cola-1; Cola2; Cola-2; COLIA1; collagen; collagen 1 alpha 2 chain; collagen a2; collagen alpha 1 chain type I; collagen alpha 2(I) chain precursor; collagen alpha-1 chain; collagen alpha-1 chain precursor; collagen alpha-1(I) chain; collagen alpha-1(I) chain preproprotein; collagen alpha-2; collagen alpha-2 chain precursor; collagen alpha-2(I) chain; collagen COL1A2; collagen I alpha-2 polypeptide; collagen I, alpha-2 polypeptide; collagen of skin, tendon and bone, alpha-1 chain; collagen of skin, tendon and bone, alpha-2 chain; collagen type I alpha 1; collagen type I alpha 1 chain; collagen type I alpha 2 chain; collagen, type 1, alpha 1; collagen, type I, alpha 1; collagen, type I, alpha 2; EDSC; Mov13; Mov-13; OI1; OI2; OI3; OI4; oim; osteogenesis imperfecta; precollagen alpha-2; prepro-alpha-1 collagen type I; pro-alpha-1 collagen type 1; procollagen type I, alpha 1; procollagen, type 1, alpha 1; procollagen, type I, alpha 1; procollagen, type I, alpha 2; similar to pro alpha 1(I) collagen; Type I Collagen; type I collagen alpha 1 chain; type I collagen alpha1; type I collagen alpha-2 chain; type I collagen pre-pro-alpha1(I) chain; type I proalpha 1; type I procollagen; type I procollagen alpha 1 chain; type I-alpha 1 collagen |
| Gene Symbols | COL1A1, COL1A2 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.