missing translation for 'onlineSavingsMsg'
Learn More

JG Finneran Associates™ Clear Polytetrafluoroethylene Snap Cap

Product Code. 10025560
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 1000
Product Code. Color unitSize
10025560 Clear Pack of 1000
1 options
This item is not returnable. View return policy

Product Code. 10025560

Brand: JG Finneran Associates™ 521011

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Diameter (Metric) Cap 11 mm
Color Clear
Material PTFE
For Use With (Application) Used on 12x32mm Snap Seal™ vials.
Quantity 1000 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.